Gene Transcripts
- Gene Symbol
- NHEJ1
- Entrez Gene ID
- 79840
- Gene Name
- non-homologous end joining factor 1
- Gene Aliases
- XLF
- Chromosome Location
- Chr. 2: 219075324 - 219160865 on Build GRCh38
- UniGene ID
- Hs.225988
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI830639.1 |
- |
6 |
669 |
|
Chr. 2 - 219077276 - 219160531 [-] |
GenBankmRNA |
BQ071822.1 |
- |
7 |
806 |
|
Chr. 2 - 219077285 - 219160798 [-] |
GenBankmRNA |
DA542708.1 |
- |
6 |
756 |
|
Chr. 2 - 219078082 - 219160830 [-] |
GenBankmRNA |
BG696101.1 |
- |
5 |
645 |
|
Chr. 2 - 219077246 - 219158360 [-] |
GenBankmRNA |
BQ645957.1 |
- |
6 |
787 |
|
Chr. 2 - 219077315 - 219160845 [-] |
View Full Product Details