Gene Transcripts
- Gene Symbol
- MAP2K3
- Entrez Gene ID
- 5606
- Gene Name
- mitogen-activated protein kinase kinase 3
- Gene Aliases
- MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
- Chromosome Location
- Chr. 17: 21284656 - 21315240 on Build GRCh38
- UniGene ID
- Hs.514012
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001316332.1 |
NP_001303261.1
|
10, 11 |
1162 |
|
NM_002756.4 |
NP_002747.2
|
9, 10 |
779 |
|
NM_145109.2 |
NP_659731.1
|
9, 10 |
1037 |
|
XM_005256723.2 |
XP_005256780.1
|
10, 11 |
1077 |
|
XM_011523959.1 |
XP_011522261.1
|
11, 12 |
1044 |
|
XM_017024857.1 |
XP_016880346.1
|
9, 10 |
906 |
|
XM_017024858.1 |
XP_016880347.1
|
10, 11 |
894 |
|
XM_017024859.1 |
XP_016880348.1
|
11, 12 |
1234 |
|
GenBank mRNA |
AB451226.1 |
- |
9, 10 |
772 |
|
AB451349.1 |
- |
9, 10 |
772 |
|
AK093838.1 |
- |
10, 11 |
1081 |
|
AK223490.1 |
- |
9, 10 |
912 |
|
AK315505.1 |
- |
9, 10 |
982 |
|
BC032478.1 |
- |
9, 10 |
972 |
5215093 |
CR536514.1 |
- |
9, 10 |
772 |
|
CR541793.1 |
- |
9, 10 |
772 |
|
D87116.1 |
- |
9, 10 |
829 |
|
GQ225578.1 |
- |
9, 10 |
772 |
|
L36719.1 |
- |
11, 12 |
1022 |
|
U66839.1 |
- |
9, 10 |
898 |
|
- Gene Symbol
- LOC100996792
- Entrez Gene ID
- 100996792
- Gene Name
- dual specificity mitogen-activated protein kinase kinase 3
- Gene Aliases
- Chromosome Location
- Chr. 17: 21772912 - 21783444 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details