Gene Transcripts

Gene Symbol
NCK2
Entrez Gene ID
8440
Gene Name
NCK adaptor protein 2
Gene Aliases
GRB4, NCKbeta
Chromosome Location
Chr. 2: 105744649 - 105894274 on Build GRCh38
UniGene ID
Hs.529244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004720.2 NP_001004720.1 4 2282
NM_001004722.3 NP_001004722.1 4 1670
NM_003581.4 NP_003572.2 5 2392
XM_006712797.3 XP_006712860.1 5 3048
XM_011511991.2 XP_011510293.1 5 2486
XM_011511992.2 XP_011510294.2 3 1614
XM_017005103.1 XP_016860592.1 6 2650
XM_017005104.1 XP_016860593.1 5 2662
XM_017005105.1 XP_016860594.1 5 2399
GenBank mRNA BC000103.2 - 4 2164
BC007195.1 - 4 2179 3608149

View Full Product Details