Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 6 | 916 | |
XM_005263438.2 | XP_005263495.1 | 6 | 923 | ||
XM_011541166.2 | XP_011539468.1 | 6 | 916 | ||
XM_017000900.1 | XP_016856389.1 | 3 | 379 | ||
XM_017000901.1 | XP_016856390.1 | 5 | 776 | ||
XM_017000902.1 | XP_016856391.1 | 6 | 916 | ||
GenBank mRNA | AB209995.1 | - | 6 | 897 | |
AK302863.1 | - | 6 | 908 | ||
AK304273.1 | - | 6 | 905 | ||
BC117166.1 | - | 5 | 813 | 40125717 | |
L34075.1 | - | 6 | 874 | ||
U88966.1 | - | 6 | 874 |