Gene Transcripts

Gene Symbol
Cpox
Entrez Gene ID
12892
Gene Name
coproporphyrinogen oxidase
Gene Aliases
CPX, Cpo, HCP, cac, nct
Chromosome Location
Chr. 16: 58670208 - 58680391 on Build GRCm38
UniGene ID
Mm.291519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007757.2 NP_031783.2 2 819
GenBank mRNA AK157277.1 - 2 702
AK166512.1 - 2 681
BC017680.1 - 2 676 4241687
CA535096.1 - 2 147
D16333.1 - 2 733

View Full Product Details