Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159289.1 | NP_001152761.1 | 4 | 913 | |
NM_001159290.1 | NP_001152762.1 | 4 | 568 | ||
NM_001242934.1 | NP_001229863.1 | 4 | 913 | ||
NM_016707.3 | NP_057916.1 | 4 | 913 | ||
GenBank mRNA | AF051525.1 | - | 4 | 851 | |
AF169037.1 | - | 4 | 851 | ||
AF186018.1 | - | 4 | 794 | ||
AK004395.1 | - | 4 | 789 | ||
AK043677.1 | - | 4 | 712 | ||
AK045556.1 | - | 4 | 569 | ||
AK140949.1 | - | 4 | 858 | ||
BC051418.1 | - | 4 | 701 |