Gene Transcripts

Gene Symbol
Nkx6-2
Entrez Gene ID
14912
Gene Name
NK6 homeobox 2
Gene Aliases
Gtx, Nkx6.2
Chromosome Location
Chr. 7: 139579376 - 139582797 on Build GRCm38
UniGene ID
Mm.28308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183248.3 NP_899071.2 3 1256
GenBank mRNA AK008173.1 - 3 1257
L08074.1 - 3 1194

View Full Product Details