Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175088.5 | NP_780297.3 | 4 | 835 | |
GenBank mRNA | AK040675.1 | - | 3 | 547 | |
AK048821.1 | - | 4 | 775 | ||
AK086117.1 | - | 5 | 838 | ||
AK134050.1 | - | 4 | 776 | ||
AK165016.1 | - | 4 | 836 | ||
AK166462.1 | - | 9 | 1611 | ||
AK172522.1 | - | 3 | 684 | ||
BC067006.1 | - | 4 | 566 |