Gene Transcripts

Gene Symbol
Tpsb2
Entrez Gene ID
17229
Gene Name
tryptase beta 2
Gene Aliases
AV011504, MMCP-6, Mcp-6, Mcp6, Mcpt6
Chromosome Location
Chr. 17: 25366303 - 25368096 on Build GRCm38
UniGene ID
Mm.7409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010781.3 NP_034911.3 3 286
GenBank mRNA BC024374.1 - 3 286
L31853.1 - 3 280
M57625.1 - 2336
M57626.1 - 4 308
X78542.1 - 280

View Full Product Details