Gene Transcripts

Gene Symbol
Mgmt
Entrez Gene ID
17314
Gene Name
O-6-methylguanine-DNA methyltransferase
Gene Aliases
AGT, AI267024, Agat
Chromosome Location
Chr. 7: 136894468 - 137128193 on Build GRCm38
UniGene ID
Mm.440219
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008598.2 NP_032624.1 4 421
GenBank mRNA AB092489.1 - 3 332
AB092490.1 - 3 332
AK090389.1 - 411
BC031888.1 - 4 390 5372377
BC096557.1 - 4 424 1366095
M84524.1 - 3 347

View Full Product Details