Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010798.2 | NP_034928.1 | 2 | 221 | |
GenBank mRNA | AK008939.1 | - | 2 | 215 | |
AK010691.1 | - | 2 | 192 | ||
AK013328.1 | - | 2 | 215 | ||
AK152211.1 | - | 2 | 202 | ||
BC024895.1 | - | 2 | 201 | 3978685 | |
BC086928.1 | - | 2 | 191 | 6762214 | |
BC100605.1 | - | 2 | 191 | 30118109 | |
L02913.1 | - | 2 | 194 | ||
L07607.1 | - | 2 | 124 | ||
L10613.1 | - | 2 | 220 | ||
Z23048.1 | - | 2 | 141 |