Gene Transcripts

Gene Symbol
Nedd1
Entrez Gene ID
17997
Gene Name
neural precursor cell expressed, developmentally down-regulated gene 1
Gene Aliases
Chromosome Location
Chr. 10: 92684745 - 92723682 on Build GRCm38
UniGene ID
Mm.2998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008682.2 NP_032708.2 14 1938
GenBank mRNA AK010591.1 - 14 1926
AK048393.1 - 14 1890
AK153746.1 - 14 1940
BC052430.1 - 14 1907 6836583
BC066870.1 - 14 1875 30476383
D10712.1 - 14 1880

View Full Product Details