Gene Transcripts

Gene Symbol
Nnmt
Entrez Gene ID
18113
Gene Name
nicotinamide N-methyltransferase
Gene Aliases
Chromosome Location
Chr. 9: 48538001 - 48607635 on Build GRCm38
UniGene ID
Mm.458542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010924.2 NP_035054.1 1 180
GenBank mRNA BC028757.1 - 1 96
CF732455.1 - 1 180
U86105.1 - 1 227
U86106.1 - 1 227
U86107.1 - 1 227
U86108.1 - 1 227
U89377.1 - 1 227

View Full Product Details