Gene Transcripts

Gene Symbol
Notch2
Entrez Gene ID
18129
Gene Name
notch 2
Gene Aliases
AI853703, N2
Chromosome Location
Chr. 3: 98013538 - 98150367 on Build GRCm38
UniGene ID
Mm.485843
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010928.2 NP_035058.2 23 4006
GenBank mRNA AK155137.1 - 5 808
AK155666.1 - 5 808
D32210.1 - 23 4000
X68279.1 - 18 2894

View Full Product Details