Gene Transcripts

Gene Symbol
Psme2
Entrez Gene ID
19188
Gene Name
proteasome (prosome, macropain) activator subunit 2 (PA28 beta)
Gene Aliases
AA589371, AI788882, PA28b
Chromosome Location
Chr. 14: 55587440 - 55591146 on Build GRCm38
UniGene ID
Mm.371573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029855.1 NP_001025026.1 8, 9 522
NM_011190.3 NP_035320.1 8, 9 622
GenBank mRNA AA267668.1 - 8, 9 513
AK012344.1 - 8, 9 551
AK088654.1 - 8, 9 549
AK143556.1 - 8, 9 554
BC005680.1 - 8, 9 542 3154255
BC057859.1 - 8, 9 619 5310526
BY498670.1 - 2, 3 128
U60329.1 - 8, 9 491

View Full Product Details