Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253904.1 | NP_001240833.1 | 39, 40 | 6249 | |
NM_001253905.1 | NP_001240834.1 | 34, 35 | 5698 | ||
NM_011424.3 | NP_035554.3 | 38, 39 | 6297 | ||
GenBank mRNA | AF113002.1 | - | 34, 35 | 5690 | |
AF125671.1 | - | 38, 39 | 6114 | ||
AK147308.1 | - | 18, 19 | 3225 | ||
AK147394.1 | - | 39, 40 | 6214 | ||
BC047524.1 | - | 2, 3 | 462 |