Gene Transcripts
- Gene Symbol
- Morf4l1
- Entrez Gene ID
- 21761
- Gene Name
- mortality factor 4 like 1
- Gene Aliases
- MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
- Chromosome Location
- Chr. 9: 90091669 - 90114820 on Build GRCm38
- UniGene ID
- Mm.426209
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039147.2 |
NP_001034236.1
|
6, 7 |
678 |
|
NM_024431.3 |
NP_077751.1
|
5, 6 |
561 |
|
GenBank mRNA |
AB042241.1 |
- |
5, 6 |
410 |
|
AF319620.1 |
- |
5, 6 |
409 |
|
AF319621.1 |
- |
6, 7 |
526 |
|
AK010528.1 |
- |
5, 6 |
330 |
|
AK011249.1 |
- |
5, 6 |
438 |
|
AK011441.1 |
- |
5, 6 |
333 |
|
AK016414.1 |
- |
5, 6 |
528 |
|
AK146210.1 |
- |
5, 6 |
396 |
|
AK146785.1 |
- |
4, 5 |
291 |
|
AK150925.1 |
- |
5, 6 |
362 |
|
AK167079.1 |
- |
5, 6 |
320 |
|
AK168327.1 |
- |
5, 6 |
396 |
|
AK168585.1 |
- |
5, 6 |
341 |
|
AK168608.1 |
- |
5, 6 |
343 |
|
AK168674.1 |
- |
5, 6 |
392 |
|
AK169065.1 |
- |
5, 6 |
393 |
|
AK181499.1 |
- |
1, 2 |
20 |
|
AK208721.1 |
- |
1, 2 |
20 |
|
BC003894.1 |
- |
5, 6 |
413 |
3593982 |
BC017619.1 |
- |
5, 6 |
413 |
4923923 |
BC083094.1 |
- |
5, 6 |
346 |
|
BC083118.1 |
- |
5, 6 |
422 |
5371036 |
BC085103.1 |
- |
6, 7 |
528 |
6490291 |
BC103783.1 |
- |
5, 6 |
425 |
6529024 |
View Full Product Details