Gene Transcripts

Gene Symbol
Pappa2
Entrez Gene ID
23850
Gene Name
pappalysin 2
Gene Aliases
PAPP-A2, PLAC3, Pappe
Chromosome Location
Chr. 1: 158711727 - 158980523 on Build GRCm38
UniGene ID
Mm.458621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085376.2 NP_001078845.1 13 3954
GenBank mRNA AK084006.1 - 13 3471
BC094560.1 - 3 214

View Full Product Details