Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011908.2 | NP_036038.1 | 3 | 984 | |
GenBank mRNA | AF044222.1 | - | 3 | 926 | |
AF044223.1 | - | 3 | 245 | ||
AK134598.1 | - | 3 | 352 | ||
AK145952.1 | - | 3 | 663 | ||
BC024507.1 | - | 5 | 834 | 4983604 | |
BC025595.1 | - | 3 | 639 | 5342735 | |
BC043729.1 | - | 5 | 834 | 4983604 |