Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011971.4 | NP_036101.1 | 2 | 197 | |
GenBank mRNA | AF060092.1 | - | 2 | 159 | |
AK002331.1 | - | 2 | 198 | ||
AK010488.1 | - | 2 | 178 | ||
AK050108.1 | - | 2 | 195 | ||
AK153162.1 | - | 2 | 230 | ||
BC014783.1 | - | 2 | 164 | 3500033 | |
BC098233.1 | - | 1 | 159 | 1547611 |