Gene Transcripts

Gene Symbol
PRCP
Entrez Gene ID
5547
Gene Name
prolylcarboxypeptidase
Gene Aliases
HUMPCP, PCP
Chromosome Location
Chr. 11: 82822940 - 82901740 on Build GRCh38
UniGene ID
Hs.523936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319214.1 NP_001306143.1 7 991
NM_005040.3 NP_005031.1 8 1353
NM_199418.3 NP_955450.2 9 1416
XM_005274093.1 XP_005274150.1 8 1083
GenBank mRNA AB451270.1 - 8 1152
AB451397.1 - 8 1152
AK090847.1 - 2 1649
AK091786.1 - 9 1243
AK302386.1 - 7 942
AK312919.1 - 8 1174
BC001500.2 - 8 1171 3510081
L13977.1 - 8 1181

View Full Product Details