Gene Transcripts
- Gene Symbol
- Rufy3
- Entrez Gene ID
- 52822
- Gene Name
- RUN and FYVE domain containing 3
- Gene Aliases
- 2810428M05Rik, 6330416M07Rik, AW455998, AW538594, D5Bwg0860e, Ripx, Rpipx
- Chromosome Location
- Chr. 5: 88564780 - 88651417 on Build GRCm38
- UniGene ID
- Mm.270469
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289774.1 |
NP_001276703.1
|
9, 10 |
1245 |
|
NM_001289775.1 |
NP_001276704.1
|
9, 10 |
1245 |
|
NM_001289776.1 |
NP_001276705.1
|
10, 11 |
1667 |
|
NM_001289777.1 |
NP_001276706.1
|
11, 12 |
1492 |
|
NM_027530.3 |
NP_081806.1
|
9, 10 |
1613 |
|
GenBank mRNA |
AK013185.1 |
- |
11, 12 |
1493 |
|
AK018184.1 |
- |
9, 10 |
1591 |
|
AK083242.1 |
- |
9, 10 |
1548 |
|
AK148795.1 |
- |
9, 10 |
909 |
|
AK154941.1 |
- |
9, 10 |
1246 |
|
AK160820.1 |
- |
9, 10 |
1247 |
|
BC017648.1 |
- |
9, 10 |
1002 |
4527858 |
BC049127.1 |
- |
9, 10 |
1552 |
6821567 |
BC058259.1 |
- |
10, 11 |
1386 |
6469137 |
CT010404.1 |
- |
9, 10 |
986 |
|
View Full Product Details