Gene Transcripts

Gene Symbol
Rfx5
Entrez Gene ID
53970
Gene Name
regulatory factor X, 5 (influences HLA class II expression)
Gene Aliases
Chromosome Location
Chr. 3: 94951957 - 94961561 on Build GRCm38
UniGene ID
Mm.24308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017395.2 NP_059091.2 9 1521
GenBank mRNA AB221679.1 - 9 1502
AF209854.1 - 9 1502
AK088686.1 - 9 1687
AK156195.1 - 1 145
BC051965.1 - 10 1558 5702640
BC057336.1 - 9 1911
BC075717.1 - 10 1994
BC118948.2 - 9 1502 40092396
BC118949.2 - 9 1502 40092397

View Full Product Details