Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009935.2 | NP_001009935.1 | 4 | 796 | |
NM_023719.2 | NP_076208.2 | 4 | 793 | ||
GenBank mRNA | AF282826.1 | - | 4 | 514 | |
AK004653.2 | - | 4 | 793 | ||
AK082476.1 | - | 4 | 791 | ||
AK088870.1 | - | 4 | 791 | ||
AK089304.1 | - | 4 | 796 | ||
AK089351.1 | - | 4 | 794 | ||
AK089403.1 | - | 4 | 796 | ||
AK155517.1 | - | 4 | 794 | ||
AK171205.1 | - | 4 | 794 | ||
AK171312.1 | - | 4 | 794 | ||
AK171425.1 | - | 4 | 794 | ||
AK171456.1 | - | 4 | 791 | ||
AK171464.1 | - | 4 | 794 | ||
AK171527.1 | - | 4 | 794 | ||
AK192038.1 | - | 4 | 391 | ||
AK214578.1 | - | 4 | 368 | ||
BC011212.1 | - | 4 | 772 | 4193517 | |
BC031850.1 | - | 4 | 771 | 3591421 |