Gene Transcripts

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
Chromosome Location
Chr. 18: 23689443 - 23955066 on Build GRCh38
UniGene ID
Hs.436367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000227.4 NP_000218.3 38 5117
NM_001127717.2 NP_001121189.2 74 9992
NM_001127718.2 NP_001121190.2 37 4949
NM_198129.2 NP_937762.2 75 10160
XM_011525978.2 XP_011524280.1 75 10187
XM_011525979.2 XP_011524281.1 75 10178
XM_011525980.2 XP_011524282.1 75 10169
XM_011525981.2 XP_011524283.1 74 10055
XM_011525982.2 XP_011524284.1 73 9890
XM_017025743.1 XP_016881232.1 58 7871
XM_017025744.1 XP_016881233.1 48 6552
GenBank mRNA AB107369.1 - 75 9919
AB208853.1 - 30 4145
AY327114.1 - 37 4924
AY327115.1 - 75 9919
AY327116.1 - 74 9751
BC043618.1 - 32 4497
L34155.1 - 38 5059
X84900.1 - 40 5338

View Full Product Details