Gene Transcripts

Gene Symbol
Yeats4
Entrez Gene ID
64050
Gene Name
YEATS domain containing 4
Gene Aliases
4930573H17Rik, B230215M10Rik, GAS41, NuBI-1
Chromosome Location
Chr. 10: 117215141 - 117224507 on Build GRCm38
UniGene ID
Mm.233529
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026570.4 NP_080846.1 7 813
GenBank mRNA AK002839.1 - 6 461
AK010522.1 - 7 814
AK012096.1 - 7 780
AK016290.1 - 2 249
AK081867.1 - 7 824
BC020043.1 - 7 781 4013548

View Full Product Details