Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025882.3 | NP_080158.1 | 1, 2 | 251 | |
GenBank mRNA | AK010135.1 | - | 1, 2 | 246 | |
AK010300.1 | - | 1, 2 | 252 | ||
AK011595.1 | - | 1, 2 | 263 | ||
AK145365.1 | - | 1, 2 | 257 | ||
AK160582.1 | - | 1, 2 | 263 | ||
AK167631.1 | - | 1, 2 | 241 | ||
AK182530.1 | - | 11 | |||
BC023189.1 | - | 1, 2 | 229 | 4952949 |