Gene Transcripts

Gene Symbol
Nudt7
Entrez Gene ID
67528
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 7
Gene Aliases
1300007B24Rik, 2210404C19Rik
Chromosome Location
Chr. 8: 114133574 - 114154739 on Build GRCm38
UniGene ID
Mm.27889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290180.1 NP_001277109.1 3 345
NM_001290181.1 NP_001277110.1 4 2239
NM_024437.4 NP_077757.2 4 499
NM_024446.5 NP_077766.3 4 439
GenBank mRNA AA616044.1 - 3 341
AF338424.1 - 4 459
AK004924.1 - 4 2195
AK011172.1 - 4 373
AK088191.1 - 3 347
AK218756.1 - 1 69
BC069843.1 - 4 455 5102473
CA460240.1 - 4 531

View Full Product Details