Gene Transcripts

Gene Symbol
Pllp
Entrez Gene ID
67801
Gene Name
plasma membrane proteolipid
Gene Aliases
0610010I06Rik, AV001002, Plapi, Tm4sf11
Chromosome Location
Chr. 8: 94674895 - 94696242 on Build GRCm38
UniGene ID
Mm.279977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026385.4 NP_080661.1 2 283
GenBank mRNA AK002477.1 - 2 284
AK018198.1 - 2 272
AK032540.1 - 2 272
BC024534.1 - 2 274 4987882

View Full Product Details