Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300804.1 | NP_001287733.1 | 6 | 1332 | |
NM_001300805.1 | NP_001287734.1 | 4 | 1012 | ||
NM_001300806.1 | NP_001287735.1 | 4 | 1281 | ||
NM_005586.3 | NP_005577.1 | 5 | 1195 | ||
XM_005249117.3 | XP_005249174.1 | 5 | 1198 | ||
XM_011514625.2 | XP_011512927.1 | 6 | 1483 | ||
XM_011514626.2 | XP_011512928.1 | 5 | 1306 | ||
XM_017010867.1 | XP_016866356.1 | 4 | 1854 | ||
GenBank mRNA | AK290735.1 | - | 4 | 1279 | |
BC007836.2 | - | 5 | 1144 | 4110841 | |
U78313.1 | - | 5 | 1137 |