Gene Transcripts

Gene Symbol
Adh6a
Entrez Gene ID
69117
Gene Name
alcohol dehydrogenase 6A (class V)
Gene Aliases
1810009C16Rik, Adh5a
Chromosome Location
Chr. 3: 138313286 - 138331134 on Build GRCm38
UniGene ID
Mm.46265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026945.1 NP_081221.1 5 568
GenBank mRNA AK007397.1 - 5 564
AK028114.1 - 5 570
BC130016.1 - 5 537 40140468

View Full Product Details