Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243584.1 | NP_001230513.1 | 6 | 754 | |
NM_001243586.1 | NP_001230515.1 | 5 | 981 | ||
NM_001243587.1 | NP_001230516.1 | 6 | 697 | ||
NM_027162.4 | NP_081438.4 | 5 | 1038 | ||
GenBank mRNA | AK003708.1 | - | 815 | ||
AK010175.1 | - | 6 | 708 | ||
AK150749.1 | - | 6 | 708 | ||
BC019515.1 | - | 6 | 665 | 4460171 | |
BC026740.1 | - | 6 | 714 | 4015105 | |
BC026745.1 | - | 6 | 524 | ||
BC055812.1 | - | 5 | 1038 | 4934728 |