Gene Transcripts

Gene Symbol
MYBPH
Entrez Gene ID
4608
Gene Name
myosin binding protein H
Gene Aliases
Chromosome Location
Chr. 1: 203167811 - 203175814 on Build GRCh38
UniGene ID
Hs.927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004997.2 NP_004988.2 9 1377
GenBank mRNA BC044226.1 - 9 1377
L05606.1 - 9 1343

View Full Product Details