Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028447.3 | NP_082723.2 | 3, 4 | 659 | |
GenBank mRNA | AK004443.1 | - | 3, 4 | 653 | |
AK143537.1 | - | 3, 4 | 651 | ||
AK182391.1 | - | 1, 2 | 116 | ||
BC014817.1 | - | 3, 4 | 575 | 3709357 |