Gene Transcripts
- Gene Symbol
- NDUFS8
- Entrez Gene ID
- 4728
- Gene Name
- NADH:ubiquinone oxidoreductase core subunit S8
- Gene Aliases
- CI-23k, CI23KD, TYKY
- Chromosome Location
- Chr. 11: 68030617 - 68036647 on Build GRCh38
- UniGene ID
- Hs.90443
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002496.3 |
NP_002487.1
|
3, 4 |
232 |
|
XM_005274013.1 |
XP_005274070.1
|
3, 4 |
329 |
|
XM_005274014.2 |
XP_005274071.1
|
3, 4 |
356 |
|
XM_011545053.2 |
XP_011543355.1
|
3, 4 |
267 |
|
GenBank mRNA |
AK223114.1 |
- |
3, 4 |
222 |
|
AK302449.1 |
- |
3, 4 |
232 |
|
AK314546.1 |
- |
3, 4 |
157 |
|
BC119754.2 |
- |
3, 4 |
161 |
40119290 |
DB498707.1 |
- |
3, 4 |
218 |
|
U65579.1 |
- |
3, 4 |
155 |
|
View Full Product Details