Gene Transcripts

Gene Symbol
Med13l
Entrez Gene ID
76199
Gene Name
mediator complex subunit 13-like
Gene Aliases
2210413I17Rik, 6330591G05Rik, 9030618F05Rik, Thrap2, Trap240L, mKIAA1025
Chromosome Location
Chr. 5: 118560706 - 118765438 on Build GRCm38
UniGene ID
Mm.206238
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172424.4 NP_766012.3 2 235
GenBank mRNA AK088795.1 - 2 278
AK142391.1 - 2 250
AK154895.1 - 2 215
AY338464.1 - 2 212

View Full Product Details