Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243146.1 | NP_001230075.1 | 4 | 307 | |
NM_001243147.1 | NP_001230076.1 | 6 | 439 | ||
NM_002533.3 | NP_002524.2 | 7 | 712 | ||
NM_206840.2 | NP_996671.1 | 6 | 638 | ||
XM_011544196.1 | XP_011542498.1 | 7 | 695 | ||
XM_011544198.1 | XP_011542500.1 | 6 | 650 | ||
XM_011544199.1 | XP_011542501.1 | 6 | 604 | ||
XM_011544200.1 | XP_011542502.1 | 5 | 578 | ||
XM_017001378.1 | XP_016856867.1 | 8 | 804 | ||
XM_017001379.1 | XP_016856868.1 | 8 | 804 | ||
XM_017001380.1 | XP_016856869.1 | 8 | 804 | ||
XM_017001381.1 | XP_016856870.1 | 8 | 801 | ||
XM_017001382.1 | XP_016856871.1 | 7 | 734 | ||
XM_017001383.1 | XP_016856872.1 | 7 | 734 | ||
XM_017001384.1 | XP_016856873.1 | 8 | 804 | ||
XM_017001385.1 | XP_016856874.1 | 8 | 804 | ||
XM_017001386.1 | XP_016856875.1 | 5 | 578 | ||
GenBank mRNA | AK297070.1 | - | 7 | 671 | |
AK297396.1 | - | 4 | 258 | ||
AK298244.1 | - | 6 | 388 | ||
AK298826.1 | - | 6 | 590 | ||
AW997091.1 | - | 4 | 457 | ||
BC005897.1 | - | 7 | 671 | ||
BC012105.1 | - | 5 | 299 | 4636004 | |
BQ883523.1 | - | 2 | 233 | ||
U68140.1 | - | 7 | 706 | ||
U78772.1 | - | 6 | 619 |