Gene Transcripts

Gene Symbol
Lce1i
Entrez Gene ID
76585
Gene Name
late cornified envelope 1I
Gene Aliases
2310069N01Rik
Chromosome Location
Chr. 3: 92777210 - 92778899 on Build GRCm38
UniGene ID
Mm.291782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029667.2 NP_083943.1 2 552
GenBank mRNA AK010137.1 - 2 555

View Full Product Details