Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190807.2 | NP_001177736.1 | 1 | 248 | |
NM_001914.3 | NP_001905.1 | 1 | 248 | ||
NM_148923.3 | NP_683725.1 | 1 | 248 | ||
XM_011525835.2 | XP_011524137.1 | 1 | 212 | ||
GenBank mRNA | AB209617.1 | - | 1 | 189 | |
AK026310.1 | - | 1 | 172 | ||
BC015182.1 | - | 1 | 176 | 3912638 | |
CA771478.1 | - | 1 | 214 | ||
CB112646.1 | - | 1 | 248 | ||
CR456990.1 | - | 1 | 107 | ||
M22865.1 | - | 1 | 159 | ||
M60174.1 | - | 1 | 107 |