Gene Transcripts

Gene Symbol
RBM24
Entrez Gene ID
221662
Gene Name
RNA binding motif protein 24
Gene Aliases
RNPC6, dJ259A10.1
Chromosome Location
Chr. 6: 17281508 - 17293875 on Build GRCh38
UniGene ID
Hs.519904
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143941.1 NP_001137413.1 4 2276
NM_001143942.1 NP_001137414.1 4 2292
NM_153020.2 NP_694565.1 3 2224
XM_005248932.3 XP_005248989.1 5 2407
XM_011514387.2 XP_011512689.1 4 2193
XM_011514388.2 XP_011512690.1 4 2518
XM_011514389.2 XP_011512691.1 3 2467
GenBank mRNA AK055391.1 - 3 2223
AK095016.1 - 5 2239
AL832199.1 - 1 6272
AL833156.1 - 2 1720

View Full Product Details