Gene Transcripts

Gene Symbol
NCOA7
Entrez Gene ID
135112
Gene Name
nuclear receptor coactivator 7
Gene Aliases
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
Chromosome Location
Chr. 6: 125780948 - 125932030 on Build GRCh38
UniGene ID
Hs.171426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122842.2 NP_001116314.1 16 3433
NM_001199619.1 NP_001186548.1 17 3449
NM_001199620.1 NP_001186549.1 18 3567
NM_001199621.1 NP_001186550.1 15 3212
NM_001199622.1 NP_001186551.1 6 1061
NM_181782.4 NP_861447.3 16 3466
XM_005266822.4 XP_005266879.1 16 3496
XM_006715340.3 XP_006715403.1 16 3423
XM_011535455.2 XP_011533757.1 16 3371
XM_017010269.1 XP_016865758.1 16 3182
XM_017010270.1 XP_016865759.1 17 3395
XM_017010271.1 XP_016865760.1 16 3393
XM_017010272.1 XP_016865761.1 16 3246
XM_017010273.1 XP_016865762.1 16 3463
XM_017010274.1 XP_016865763.1 16 3552
GenBank mRNA AF493978.1 - 17 3449
AJ420542.1 - 8 1399
AK094706.1 - 6 953
AL832621.2 - 16 3226
AL832628.2 - 16 3188
AL834442.1 - 14 2854
BC033612.1 - 9 2304
BX537385.1 - 16 3261
KC238672.1 - 6 1038

View Full Product Details