Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100816.1 | NP_001094286.1 | 2 | 962 | |
NM_004270.4 | NP_004261.1 | 2 | 678 | ||
GenBank mRNA | AF031383.1 | - | 2 | 636 | |
AF104251.1 | - | 1 | 570 | ||
AK308393.1 | - | 2 | 642 | ||
BC005250.1 | - | 2 | 632 | ||
BC006968.1 | - | 2 | 632 | ||
BT007368.1 | - | 1 | 570 | ||
CR457009.1 | - | 1 | 570 | ||
CR541696.1 | - | 1 | 570 |