Gene Transcripts

Gene Symbol
SLC25A4
Entrez Gene ID
291
Gene Name
solute carrier family 25 member 4
Gene Aliases
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1
Chromosome Location
Chr. 4: 185143263 - 185150384 on Build GRCh38
UniGene ID
Hs.246506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001151.3 NP_001142.2 2 265
GenBank mRNA AB209764.1 - 1 1532
AK291902.1 - 2 259
BC008664.1 - 2 253 3871960
BC061589.1 - 2 271 6195081
BC063643.1 - 2 254 4708068
BG707401.1 - 2 292
BI544547.1 - 2 292
J02966.1 - 2 258
KU177900.1 - 2 155
KU177902.1 - 2 155

Gene Symbol
LOC101928877
Entrez Gene ID
101928877
Gene Name
uncharacterized LOC101928877
Gene Aliases
Chromosome Location
Chr. 4: 185051876 - 185107239 on Build GRCh38
UniGene ID
Hs.246506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BG772330.1 - 5 442

View Full Product Details