Gene Transcripts

Gene Symbol
MAGEH1
Entrez Gene ID
28986
Gene Name
MAGE family member H1
Gene Aliases
APR-1, APR1, MAGEH
Chromosome Location
Chr. X: 55452089 - 55453568 on Build GRCh38
UniGene ID
Hs.279819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014061.4 NP_054780.2 1 849
GenBank mRNA AF143235.4 - 1 845
AF320912.1 - 1 774
AK313527.1 - 1 773
BC011954.1 - 1 800 3880523

View Full Product Details