Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013841.1 | NP_001013863.1 | 3, 4 | 387 | |
NM_017720.2 | NP_060190.2 | 3, 4 | 387 | ||
XM_011528123.1 | XP_011526425.1 | 3, 4 | 414 | ||
GenBank mRNA | AJ245719.1 | - | 3, 4 | 348 | |
AK000241.1 | - | 3, 4 | 358 | ||
AK125774.1 | - | 5, 6 | 897 | ||
BC000795.1 | - | 3, 4 | 338 | 3451782 | |
BF677969.1 | - | 3, 4 | 387 |