Gene Transcripts

Gene Symbol
LBH
Entrez Gene ID
81606
Gene Name
limb bud and heart development
Gene Aliases
Chromosome Location
Chr. 2: 30231531 - 30260033 on Build GRCh38
UniGene ID
Hs.567598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030915.3 NP_112177.2 3 1639
XM_017005059.1 XP_016860548.1 3 1797
GenBank mRNA AF110224.1 - 1 408
AF258555.1 - 1 630
AK023425.1 - 1 706
AK027232.1 - 3 1431
AK223541.1 - 3 1575
AL136703.1 - 3 1644
BC109376.1 - 3 1643 6495944
BC126434.1 - 3 1612 8992150
BC130415.1 - 3 1611 40146446
EF025586.1 - 3 1617

View Full Product Details