Gene Transcripts

Gene Symbol
LIN54
Entrez Gene ID
132660
Gene Name
lin-54 DREAM MuvB core complex component
Gene Aliases
CXCDC1, JC8.6, MIP120, TCX1
Chromosome Location
Chr. 4: 82924603 - 83012951 on Build GRCh38
UniGene ID
Hs.96952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115007.2 NP_001108479.1 8 950
NM_001115008.2 NP_001108480.1 8 1151
NM_001288996.1 NP_001275925.1 5 1547
NM_001288997.1 NP_001275926.1 8 888
NM_194282.3 NP_919258.2 8 1814
NR_110253.1 5 1473
NR_110254.1 7 1147
XM_005262750.4 XP_005262807.1 7 1623
XM_006714081.3 XP_006714144.1 7 1549
XM_017007728.1 XP_016863217.1 5 1356
XM_017007729.1 XP_016863218.1 6 1539
XM_017007730.1 XP_016863219.1 6 928
GenBank mRNA AB111889.1 - 7 1134
AK292769.1 - 8 898
AK294256.1 - 8 1743
AK299787.1 - 3 356
BC109277.2 - 5 1349
BC109278.2 - 5 1422 40028371
BX537919.1 - 7 1048
BX640657.1 - 8 877
BX640966.1 - 8 1809

View Full Product Details