Gene Transcripts

Gene Symbol
MARCKSL1
Entrez Gene ID
65108
Gene Name
MARCKS like 1
Gene Aliases
F52, MACMARCKS, MLP, MLP1, MRP
Chromosome Location
Chr. 1: 32333829 - 32336239 on Build GRCh38
UniGene ID
Hs.75061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023009.6 NP_075385.1 2 1391
NR_052852.1 2 1166
GenBank mRNA AL713653.1 - 2 1374
AY007131.1 - 2 1374
BC007904.2 - 2 1376 4138092
BC066915.1 - 2 1385 5259353
X70326.1 - 2 1198

View Full Product Details