Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281746.1 | NP_001268675.1 | 3 | 394 | |
NM_001281747.1 | NP_001268676.1 | 3 | 376 | ||
NM_138569.2 | NP_612636.2 | 3 | 394 | ||
XM_005249476.4 | XP_005249533.1 | 3 | 347 | ||
XM_005249478.4 | XP_005249535.1 | 2 | 201 | ||
XM_005249480.4 | XP_005249537.1 | 3 | 347 | ||
XM_006715245.3 | XP_006715308.2 | 3 | 486 | ||
XM_017011465.1 | XP_016866954.1 | 3 | 483 | ||
XM_017011466.1 | XP_016866955.1 | 3 | 480 | ||
XM_017011467.1 | XP_016866956.1 | 3 | 489 | ||
XM_017011468.1 | XP_016866957.1 | 3 | 488 | ||
XM_017011469.1 | XP_016866958.1 | 3 | 280 | ||
XM_017011470.1 | XP_016866959.1 | 3 | 578 | ||
GenBank mRNA | AK055530.1 | - | 3 | 319 | |
AK294797.1 | - | 2 | 191 | ||
AK294890.1 | - | 3 | 347 | ||
AK311190.1 | - | 3 | 347 | ||
AK316157.1 | - | 3 | 327 | ||
BC009010.1 | - | 2 | 141 | 4158300 | |
BX091237.1 | - | 3 | 261 | ||
BX647567.1 | - | 3 | 340 | ||
DA761851.1 | - | 3 | 394 | ||
DC322019.1 | - | 2 | 220 | ||
KU178822.1 | - | 2 | 95 | ||
KU178823.1 | - | 3 | 251 | ||
KU178824.1 | - | 3 | 251 | ||
KU178825.1 | - | 3 | 251 | ||
KU178826.1 | - | 3 | 251 |