Gene Transcripts

Gene Symbol
MYOM2
Entrez Gene ID
9172
Gene Name
myomesin 2
Gene Aliases
TTNAP
Chromosome Location
Chr. 8: 2045043 - 2145456 on Build GRCh38
UniGene ID
Hs.443683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003970.3 NP_003961.3 4 521
XM_006716237.1 XP_006716300.1 4 486
GenBank mRNA BC052969.1 - 4 474
X69089.1 - 4 431

View Full Product Details